Skip to Content
Merck
All Photos(1)

Documents

W519103

Sigma-Aldrich

Methyl formate

≥95%

Synonym(s):

Formic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2CH3
CAS Number:
Molecular Weight:
60.05
Beilstein:
1734623
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.642
NACRES:
NA.21

biological source

synthetic

vapor density

2.1 (vs air)

Assay

≥95%

form

liquid

autoignition temp.

842 °F

expl. lim.

23 %

refractive index

n20/D 1.343 (lit.)

pH

4.0-5.0 (20 °C, 200 g/L)

bp

32-34 °C (lit.)

mp

−100 °C (lit.)

density

0.974 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; plum

SMILES string

[H]C(=O)OC

InChI

1S/C2H4O2/c1-4-2-3/h2H,1H3

InChI key

TZIHFWKZFHZASV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl formate finds application as a fumigant in raisins and dried currants.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 1 - STOT SE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Winter R
A Consumer's Dictionary of Food Additives, 352-352 null
Hiroya Yurimoto et al.
Yeast (Chichester, England), 21(4), 341-350 (2004-03-26)
Methyl formate synthesis during growth on methanol by methylotrophic yeasts has been considered to play a role in formaldehyde detoxification. An enzyme that catalyses methyl formate synthesis was purified from methylotrophic yeasts, and was suggested to belong to a family
Alexandre Faure et al.
The Journal of chemical physics, 135(2), 024301-024301 (2011-07-20)
A potential energy surface for helium interacting with methyl formate has been computed using high-level electronic structure methods. The interaction energies obtained on a three-dimensional grid have been fitted by an analytic function of interatomic distances with correct asymptotic behavior
A Nihlén et al.
International archives of occupational and environmental health, 73(7), 479-487 (2000-11-01)
A toxicokinetic (TK) model was developed to describe the inhalation exposure in humans to methyl formate (MF), a catalyst used in foundries, and to discuss biological monitoring. The TK model consisted of four compartments: MF, the metabolites--methanol (MeOH) and formic
Shigeo Kondo et al.
Journal of hazardous materials, 138(1), 1-8 (2006-06-20)
Carbon dioxide dilution effect on the flammability limits was measured for various flammable gases. The obtained values were analyzed using the extended Le Chatelier's formula developed in a previous study. As a result, it has been found that the flammability

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service