Merck
  • Alcohol dehydrogenases that catalyse methyl formate synthesis participate in formaldehyde detoxification in the methylotrophic yeast Candida boidinii.
Alcohol dehydrogenases that catalyse methyl formate synthesis participate in formaldehyde detoxification in the methylotrophic yeast Candida boidinii.
Yeast (Chichester, England) (2004-03-26)
Hiroya Yurimoto, Bumjun Lee, Fumi Yasuda, Yasuyoshi Sakai, Nobuo Kato
ABSTRACT
Methyl formate synthesis during growth on methanol by methylotrophic yeasts has been considered to play a role in formaldehyde detoxification. An enzyme that catalyses methyl formate synthesis was purified from methylotrophic yeasts, and was suggested to belong to a family of alcohol dehydrogenases (ADHs). In this study we report the gene cloning and gene disruption analysis of three ADH-encoding genes in the methylotrophic yeast Candida boidinii (CbADH1, CbADH2 and CbADH3) in order to clarify the physiological role of methyl formate synthesis. From the primary structures of these three genes, CbAdh1 was shown to be cytosolic and CbAdh2 and CbAdh3 were mitochondrial enzymes. Gene products of CbADH1, CbADH2 and CbADH3 expressed in Escherichia coli showed both ADH- and methyl formate-synthesizing activities. The results of gene-disruption analyses suggested that methyl formate synthesis was mainly catalysed by a cytosolic ADH (CbAdh1), and this enzyme contributed to formaldehyde detoxification through glutathione-independent formaldehyde oxidation during growth on methanol by methylotrophic yeasts.
Sigma-Aldrich
Methyl formate, spectrophotometric grade, ≥98%
Sigma-Aldrich
Methyl formate, reagent grade, 97%
Sigma-Aldrich
Methyl formate, anhydrous, 99%
Supelco
Methyl formate, analytical standard