Skip to Content
Merck
All Photos(2)

Documents

112682

Sigma-Aldrich

Ethyl formate

reagent grade, 97%

Synonym(s):

Formic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOC2H5
CAS Number:
Molecular Weight:
74.08
Beilstein:
906769
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39022251
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

vapor density

2.5 (vs air)

vapor pressure

15.16 psi ( 55 °C)
3.79 psi ( 20 °C)

Assay

97%

autoignition temp.

851 °F

expl. lim.

16 %

refractive index

n20/D 1.359 (lit.)

bp

52-54 °C (lit.)

mp

−80 °C (lit.)

density

0.921 g/mL at 20 °C (lit.)

SMILES string

CCOC=O

InChI

1S/C3H6O2/c1-2-5-3-4/h3H,2H2,1H3

InChI key

WBJINCZRORDGAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Ethyl formate is an ethyl ester of formic acid. Its microwave spectrum indicates the presence of two isomeric forms having the ethyl group cis to the carbonyl oxygen atom but having different arrangement of the methyl group about the -CH2-O bond. Kinetics of its pyrolytic decomposition to the corresponding acid and alkene has been reported.

Application

Ethyl formate has been used in the preparation of N-(2-triethoxysilylpropyl) formamide. It has been used in the synthesis of N-methyl-3-phenylbicyclo[2.2.1]heptan-2-amine hydrochloride.
It may be used in the synthesis of isoflavone.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave spectrum and rotational isomerism of ethyl formate.
Riveros JM and Wilson Jr EB.
J. Chem. Phys. , 46(12), 4605-4612 (1967)
Tristan F Kinde et al.
Analytical chemistry, 87(5), 2702-2709 (2015-02-11)
While the use of sodium dodecyl sulfate (SDS) in separation buffers allows efficient analysis of complex mixtures, its presence in the sample matrix is known to severely interfere with the mass-spectrometric characterization of analyte molecules. In this article, we report
Eagleson M.
Concise Encyclopedia Chemistry, 413-413 (1994)
The kinetics of the pyrolysis of ethyl and isopropyl formates and acetates.
Blades AT.
Canadian Journal of Chemistry, 32(4), 366-372 (1954)
Hanjin Im et al.
Bioresource technology, 193, 386-392 (2015-07-06)
This work addresses an unprecedented way of co-producing biodiesel (FAEE) and valuable chemicals of ethyl levulinate (EL), ethyl formate (EF) and diethyl ether (DEE) from wet in situ transesterification of microalgae. EL, EF, and DEE were significantly produced up to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service