Skip to Content
Merck
All Photos(1)

Documents

W343502

Sigma-Aldrich

3-Methyl-2-cyclopenten-1-one

97%, FG

Synonym(s):

3-Methyl-2-cyclopentenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C5H5(=O)
CAS Number:
Molecular Weight:
96.13
FEMA Number:
3435
Beilstein:
1280476
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.112
NACRES:
NA.21

biological source

synthetic

grade

FG

reg. compliance

EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 110

Assay

97%

refractive index

n20/D 1.488 (lit.)

bp

74 °C/15 mmHg (lit.)

mp

3-5 °C (lit.)

density

0.980 g/mL at 20 °C
0.971 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fruity; sweet

SMILES string

CC1=CC(=O)CC1

InChI

1S/C6H8O/c1-5-2-3-6(7)4-5/h4H,2-3H2,1H3

InChI key

CHCCBPDEADMNCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methyl-2-cyclopenten-1-one is a component of smoke flavoring.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Study of the components of a solid smoke flavouring preparation.
Guillen MD & Manzanos MJ.
Food Chemistry, 55(3), 251-257 (1996)
Isolation and identification of the components of the tar of hickory wood smoke.
Hruzan DE, et al.
Journal of Agricultural and Food Chemistry, 22(1), 123-126 (1974)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service