Skip to Content
Merck
All Photos(2)

Key Documents

165131

Sigma-Aldrich

2,5-Hexanedione

≥98%

Synonym(s):

Acetonylacetone

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$122.00
100 G
$323.00

$122.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$122.00
100 G
$323.00

About This Item

Linear Formula:
CH3COCH2CH2COCH3
CAS Number:
Molecular Weight:
114.14
Beilstein:
506525
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$122.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor pressure

0.43 mmHg ( 20 °C)

Assay

≥98%

form

liquid

refractive index

n20/D 1.425 (lit.)

bp

191 °C (lit.)

mp

−6-−5 °C (lit.)

solubility

alcohol: miscible
diethyl ether: miscible
water: miscible

density

0.973 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC(=O)CCC(C)=O

InChI

1S/C6H10O2/c1-5(7)3-4-6(2)8/h3-4H2,1-2H3

InChI key

OJVAMHKKJGICOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,5-Hexanedione is a major metabolite of the neurotoxic industrial solvent methyl n-butyl ketone[1]. It reacts with amines to yield 2,5-dimethylpyrroles[2].

Application

2,5-Hexanedione was used as starting reagent in the synthesis of trans-2,5-dimethylpyrrolidine[3].
It was used as reference standard for the determination of 2,5HD in human urine using gas chromatography-electron capture detection and gas chromatography-mass selective detection.[4]

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2

Target Organs

Nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E K Allard et al.
Toxicology and applied pharmacology, 137(2), 149-156 (1996-04-01)
The Sertoli cell toxicant 2,5-hexanedione (2,5-HD) causes irreversible testicular atrophy in rats. After toxicant exposure, only Sertoli cells, stem cells, and a few spermatogonia remain in the seminiferous epithelium. In this study, the number, type, and fate of the remaining
D C Anthony et al.
Toxicology and applied pharmacology, 71(3), 372-382 (1983-12-01)
3,4-Dimethyl-2,5-hexanedione and 2,5-hexanedione were reacted with model amines to yield N-substituted 2,3,4,5-tetramethylpyrroles and 2,5-dimethylpyrroles, respectively. When compared to the unsubstituted parent compound 2,5-hexanedione, 3,4-dimethyl-2,5-hexanedione was found to cyclize approximately eight times as rapidly on a molar basis at 37 degrees
An improved synthesis of (-)-(2R, 5R)-2, 5-dimethylpyrrolidine.
Short RP, et al.
The Journal of Organic Chemistry, 54(7), 1755-1756 (1989)
Gas chromatographic method for the sensitive determination of 2, 5-hexanedione using electron capture and mass-selective detection.
Konidari, C. N., C. D. Stalikas, and M. I. Karayannis
Analytica Chimica Acta, 442.2, 231-239 (2001)
Sarah N Campion et al.
Toxicological sciences : an official journal of the Society of Toxicology, 117(2), 457-465 (2010-07-10)
Sertoli cells are essential for testicular germ cell maintenance and survival. We made the unexpected observation that x-radiation (x-ray)-induced germ cell loss is attenuated by co-exposure with the Sertoli cell toxicant 2,5-hexanedione (HD). The mechanisms underlying this attenuation of germ

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service