W270015
Methyl cyclopentenolone
anhydrous, natural, 97%, FG
Synonym(s):
Cyclotene
Select a Size
$58.50
Select a Size
About This Item
$58.50
Recommended Products
grade
FG
Fragrance grade
Halal
Kosher
anhydrous
natural
Agency
follows IFRA guidelines
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515
Assay
97%
greener alternative product characteristics
Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
mp
104-108 °C
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
greener alternative category
Organoleptic
bread; burnt; caramel; maple; roasted; sweet
SMILES string
CC1CCC(=O)C1=O
InChI
1S/C6H8O2/c1-4-2-3-5(7)6(4)8/h8H,2-3H2,1H3
InChI key
CFAKWWQIUFSQFU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
212.0 °F
Flash Point(C)
100 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service