Skip to Content
Merck
All Photos(1)

Key Documents

W315230

Sigma-Aldrich

Ethylcyclopentenolone

natural, 97%, FG

Synonym(s):

3-Ethylcyclopentan-1,2-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O2
CAS Number:
Molecular Weight:
126.15
FEMA Number:
3152
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
07.057
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

97%

refractive index

n/D 1.476

density

1.066 at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

fragrance allergen

no known allergens

Organoleptic

caramel; maple; sweet

storage temp.

2-8°C

SMILES string

O=C1CCC(CC)=C1O

InChI

1S/C7H10O2/c1-2-5-3-4-6(8)7(5)9/h9H,2-4H2,1H3

InChI key

JHWFWLUAUPZUCP-UHFFFAOYSA-N

Caution

Air Sensitive (Nitrogen flush and store all ppk and bulk under nitrogen)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service