Skip to Content
Merck
All Photos(1)

Key Documents

W376108

Sigma-Aldrich

5-Methyl-2-hepten-4-one, predominantly trans

98%, stabilized, FG

Synonym(s):

Hazelnut ketone

Sign Into View Organizational & Contract Pricing

Select a Size

W376108-SAMPLE-K
$54.60
100 G
$535.00
1 KG
$2,760.00

$54.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W376108-SAMPLE-K
$54.60
100 G
$535.00
1 KG
$2,760.00

About This Item

Linear Formula:
C2H5CH(CH3)COCH=CHCH3
CAS Number:
Molecular Weight:
126.20
FEMA Number:
3761
Council of Europe no.:
2085
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.139
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$54.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

Assay

98%

may contain

<0.50% alpha-tocopherol, synthetic as stabilizer

refractive index

n20/D 1.444 (lit.)

bp

170 °C (lit.)

density

0.845 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

hazelnut; pungent; nutty

SMILES string

CCC(C)C(/C=C/C)=O

InChI

1S/C8H14O/c1-4-6-8(9)7(3)5-2/h4,6-7H,5H2,1-3H3/b6-4+

InChI key

ARJWAURHQDJJAC-GQCTYLIASA-N

Related Categories

General description

5-Methyl-2-hepten-4-one predominantly trans is the characteristic aroma compound of hazelnuts.[1][113}

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

141.8 °F - closed cup

Flash Point(C)

61 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation, synthesis, and absolute configuration of filbertone?the principal flavor component of the hazelnut
Jauch J, et al.
Angewandte Chemie (International Edition in English), 28(8), 1022-1023 (1989)
Filbertone as a Marker for the Assessment of Hazelnut Spread Quality.
Cizkova H, et al.
Czech Journal of Food Sciences, 31(1) (2013)
Youna Moon et al.
Nutrients, 11(8) (2019-08-02)
The aim of this research was to estimate the preventive effects of filbertone, the main flavor compound in hazelnuts, on lipid accumulation in the adipose tissue of mice fed a high-fat diet (HFD) and to reveal the underlying molecular mechanisms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service