Skip to Content
Merck
All Photos(1)

Key Documents

W315214

Sigma-Aldrich

3-Ethyl-2-hydroxy-2-cyclopenten-1-one solution

50 wt. % in propylene glycol, FG

Sign Into View Organizational & Contract Pricing

Select a Size

W315214-SAMPLE-K
$54.60
1 KG
$509.00

$54.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
W315214-SAMPLE-K
$54.60
1 KG
$509.00

About This Item

FEMA Number:
3152
Council of Europe no.:
759
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.057
EC Index Number:
2446065
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

$54.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk OrderRequest more information

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 872/2012

concentration

50 wt. % in propylene glycol

refractive index

n20/D 1.475

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

caramel; maple; sweet

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

224.6 °F

Flash Point(C)

107 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service