Skip to Content
Merck
All Photos(1)

Key Documents

W288500

Sigma-Aldrich

3-Phenyl-1-propanol

≥98%, FCC

Synonym(s):

3-Phenylpropyl alcohol, Hydrocinnamyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)3OH
CAS Number:
Molecular Weight:
136.19
FEMA Number:
2885
Beilstein:
1857542
EC Number:
Council of Europe no.:
80
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.031
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

FCC
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.526 (lit.)

bp

119-121 °C/12 mmHg (lit.)

mp

−18 °C (lit.)

density

1.001 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

balsam; cinnamon; hyacinth; floral; spicy

SMILES string

OCCCc1ccccc1

InChI

1S/C9H12O/c10-8-4-7-9-5-2-1-3-6-9/h1-3,5-6,10H,4,7-8H2

InChI key

VAJVDSVGBWFCLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Phenyl-1-propanol is a volatile compound that is reported to occur in styrax gum[1], guava fruit[2], and beeswax.[3]

Application

Preservative in cosmetics - This alcohol a natural fragrance with antimicrobial properties against bacteria and molds. It is used in combination with Heliotropin or Piperonal as a preservative for cosmetic products.
This alcohol or its ester derivatives are used as fragrance component in fresh flower compositions, like lilac, hyacinth, and lily of the valley because of the balsamic odor character.

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical composition of the essential oils from Turkish and Honduras Styrax.
Fernandez X, et al.
Flavour and Fragrance Journal, 20(1), 70-73 (2005)
Jörgen Samuelsson et al.
Journal of chromatography. A, 1217(46), 7215-7221 (2010-10-12)
The elution by characteristic points (ECP) method is a very rapid and precise method for determination of the phase system equilibrium of phase systems in broad solute concentration ranges. Thus, the method is especially suitable for rapid characterization of high
S P Bhatia et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49 Suppl 2, S246-S251 (2011-08-23)
A toxicologic and dermatologic review of 3-phenyl-1-propanol when used as a fragrance ingredient is presented. 3-Phenyl-1-propanol is a member of the fragrance structural group cinnamyl phenylpropyl compounds. The common characteristic structural element of cinnamyl phenylpropyl materials is an aryl substituted
Soyeong Kang et al.
The Journal of organic chemistry, 75(1), 237-240 (2009-12-05)
A highly efficient, enantioselective sequence has been developed for the synthesis of (S)- and (R)-dapoxetine. The pathways involve the intermediacy of the 6-membered-ring sulfamate esters 4, which were generated by Du Bois asymmetric C-H amination reactions of the prochiral sulfamate
Chemical composition of the essential oil and headspace solid-phase microextraction of the guava fruit (Psidium guajava L.).
Paniandy JC, et al.
J. Essent. Oil Res., 12(2), 153-158 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service