Skip to Content
Merck
All Photos(1)

Key Documents

189685

Sigma-Aldrich

4-Phenyl-2-butanol

97%

Synonym(s):

α−Methyl-benzenepropanol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$62.70

$62.70


Estimated to ship onMarch 24, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$62.70

About This Item

Linear Formula:
C6H5CH2CH2CH(OH)CH3
CAS Number:
Molecular Weight:
150.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$62.70


Estimated to ship onMarch 24, 2025


Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.5140 (lit.)

bp

132 °C/14 mmHg (lit.)

density

0.970 g/mL at 25 °C (lit.)

functional group

hydroxyl
phenyl

SMILES string

CC(O)CCc1ccccc1

InChI

1S/C10H14O/c1-9(11)7-8-10-5-3-2-4-6-10/h2-6,9,11H,7-8H2,1H3

InChI key

GDWRKZLROIFUML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Phenyl-2-butanol undergoes (η5-pentaphenylcyclopentadienyl)RuCl(CO)2 catalyzed racemization efficiently at room temperature in the presence of a base[1]. 4-Phenyl-2-butanol on reaction with sulfuric acid yields polymer[2].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

New efficient ruthenium catalysts for racemization of alcohols at room temperature.
Csjernyik G, et al.
Tetrahedron Letters, 45(36), 6799-6802 (2004)
New Friedel-Crafts chemistry. XIX. Cyclialkylations of some phenylalkanols.
Khalaf AA and Roberts RM.
The Journal of Organic Chemistry, 34(11), 3571-3574 (1969)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service