Skip to Content
Merck
All Photos(2)

Documents

80133

Supelco

Methyl cyclopentenolone

analytical standard

Synonym(s):

Cyclotene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

104-108 °C
104-108 °C

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC1CCC(=O)C1=O

InChI

1S/C6H8O2/c1-4-2-3-5(7)6(4)8/h8H,2-3H2,1H3

InChI key

CFAKWWQIUFSQFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl cyclopentenolone is an organic compound, commonly used in commercial products such as in E-liquids for electronic cigarettes.

Application

Methyl cyclopentenolone may be used as an analytical reference standard for the quantification of the analyte in commercial products using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multicomponent analysis of replacement liquids of electronic cigarettes using chromatographic techniques
Kavvalakis.PM, et al.
Journal of Analytical Toxicology, 39(4), 262-269 (2015)
Comprehensive determination of flavouring additives and nicotine in e-cigarette refill solutions. Part I: Liquid chromatography-tandem mass spectrometry analysis.
Aszyk J, et al.
Journal of Chromatography A, 1519, 45-54 (2017)
Seo Young Choi et al.
Biochimica et biophysica acta, 1770(12), 1612-1619 (2007-10-13)
Peroxisome proliferator-activated receptors (PPARs) are members of the nuclear hormone receptor superfamily that plays a pivotal role in regulating inflammatory gene expression. The purpose of this study was to investigate the effects of coffee extract, 3-methyl-1,2-cyclopentanedione (3-MCP) on PPARs in
Jae Heun Chung et al.
Journal of agricultural and food chemistry, 55(16), 6787-6792 (2007-07-17)
Activation of the redox-sensitive transcription factor, nuclear factor-kappa B (NF-kappaB), plays a central role in inflammation and aging processes by inducing pro-inflammatory genes. The present study was designed to unravel the molecular mechanisms underlying the anti-inflammation effects of 3-methyl-1,2-cyclopentanedione (3-MCP)
Ae Ra Kim et al.
The Journal of pharmacy and pharmacology, 54(10), 1385-1392 (2002-10-25)
It has been known that reactive oxygen and nitrogen species such as nitric oxide (NO), superoxide radical (*O2-) and their byproduct peroxynitrite (ONOO-) induce cellular and tissue injury, ultimately resulting in several human diseases. In this study, we examined scavenging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service