Skip to Content
Merck
All Photos(1)

Key Documents

W201103

Sigma-Aldrich

Adipic acid

≥99.5%

Synonym(s):

Hexanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CH2)4COOH
CAS Number:
Molecular Weight:
146.14
FEMA Number:
2011
Beilstein:
1209788
EC Number:
Council of Europe no.:
26
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39021711
PubChem Substance ID:
Flavis number:
8.026
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

reg. compliance

FDA 21 CFR 117

vapor density

5 (vs air)

vapor pressure

1 mmHg ( 159.5 °C)

Assay

≥99.5%

autoignition temp.

788 °F

bp

265 °C/100 mmHg (lit.)

mp

151-154 °C (lit.)

solubility

H2O: soluble 23 g/L at 25 °C

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

OC(=O)CCCCC(O)=O

InChI

1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)

InChI key

WNLRTRBMVRJNCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

384.8 °F - closed cup

Flash Point(C)

196 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roohan Rakhshaee et al.
Journal of hazardous materials, 189(1-2), 158-166 (2011-03-15)
The removal of methylene blue (MB) as a cationic dye from aqueous solution by the stabilized Fe(3)O(4) nano-particles with the extracted pectin from apple waste (FN-PA) increased due to using the cross-linked forms of the bound pectin on the nano-particles
H V Pawar et al.
Colloids and surfaces. B, Biointerfaces, 102, 102-110 (2012-09-26)
Streptomycin (STP) and diclofenac (DLF) loaded film dressings were prepared by blending Polyox(®) (POL) with four hydrophilic polymers [hydroxypropylmethylcellulose (HPMC), carrageenan (CAR), sodium alginate (SA) or chitosan (CS)] using glycerol (GLY) as plasticiser. The films were characterised by scanning electron
Holger Noack et al.
Inorganic chemistry, 50(4), 1194-1202 (2011-01-28)
Adipic acid is a key compound in the chemical industry, where it is mainly used in the production of polymers. The conventional process of its generation requires vast amounts of energy and, moreover, produces environmentally deleterious substances. Thus, there is
J B J H van Duuren et al.
Biotechnology and bioengineering, 108(6), 1298-1306 (2011-02-18)
A limited life cycle assessment (LCA) was performed on a combined biological and chemical process for the production of adipic acid, which was compared to the traditional petrochemical process. The LCA comprises the biological conversion of the aromatic feedstocks benzoic
Krzysztof J Paluch et al.
Journal of pharmaceutical sciences, 100(8), 3268-3283 (2011-04-08)
The production of salt or cocrystalline forms is a common approach to alter the physicochemical properties of pharmaceutical compounds. The goal of this work was to evaluate the impact of anion choice (succinate, adipate, and sulfate) on the physicochemical characteristics

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service