Skip to Content
Merck
All Photos(1)

Key Documents

236365

Sigma-Aldrich

Sebacoyl chloride

99%

Synonym(s):

Sebacyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$120.00
50 G
$406.00

$120.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$120.00
50 G
$406.00

About This Item

Linear Formula:
ClCO(CH2)8COCl
CAS Number:
Molecular Weight:
239.14
Beilstein:
1365665
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$120.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.468 (lit.)

bp

168 °C/12 mmHg (lit.)

mp

−5-−3 °C (lit.)

density

1.121 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)CCCCCCCCC(Cl)=O

InChI

1S/C10H16Cl2O2/c11-9(13)7-5-3-1-2-4-6-8-10(12)14/h1-8H2

InChI key

WMPOZLHMGVKUEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sebacocyl chloride (SC) can be used as a lipophilic monomer for the formation of polyamide microcapsules by an interfacial polycondensation reaction.[1] It can also be utilized in the synthesis of modified chitosan nanoparticle hydrogels with high absorption capacity and high swelling ratio for the removal of metal ions from an aqueous solution.[2] SC can also be used in the preparation of sebacic biscyclocarbonate, which can be used in the synthesis of biobased nonisocyanate polyurethanes.[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of robust polyamide microcapsules by interfacial polycondensation of p-phenylenediamine and sebacoyl chloride and plasticization with oleic acid
Rosa N, et al.
Journal of Microencapsulation, 32(4), 349-357 (2015)
Chitosan nanoparticle hydrogel based sebacoyl moiety with remarkable capability for metal ion removal from aqueous systems
Kandile NG and Mohamed HM
International Journal of Biological Macromolecules, 122(4), 578-586 (2019)
Synthesis and in vitro Anti-tumor Activity of Some New Sebacoyl Chloride Based Heterocycles
H El Azab I and AA EN
Current Organic Synthesis, 14(2), 309-320 (2017)
D Poncelet et al.
Journal of microencapsulation, 11(1), 31-40 (1994-01-01)
A microencapsulation technique is proposed involving the formation of a polyethyleneimine (PEI) membrane crosslinked by an acid dichloride. The membranes were formed at pH 8 in a non-polar solvent, conditions which are better suited for the encapsulation of biocatalysts or
Zhenhua Xu et al.
International journal of pharmaceutics, 426(1-2), 182-192 (2012-01-24)
In this study we describe a novel polymer, mPPS-FA, synthesized as a potential gene transfer vector. To complete mPPS-FA, folic acid was conjugated to a backbone (named mPPS) consisting of a copolymer of methyl PEG-2000, PEI-600, and sebacoyl chloride. (1)H

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service