Skip to Content
Merck
All Photos(2)

Key Documents

G9284

Sigma-Aldrich

Guanidine hydrochloride solution

Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5

Synonym(s):

Aminoformamidine hydrochloride, Guanidinium chloride, Guanidium chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$597.00
500 ML
$2,230.00

$597.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$597.00
500 ML
$2,230.00

About This Item

Empirical Formula (Hill Notation):
CH5N3 · HCl
CAS Number:
Molecular Weight:
95.53
Beilstein:
3591990
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.55

$597.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Application

Guanidine hydrochloride solution has been used:
  • for protein denaturation
  • to homogenize hypothalamus, thalamus, prefrontal cortex, and brainstem samples for total β-amyloid (Aβ) extraction and Aβ42 quantification
  • as a chaotropic denaturant to treat human immunodeficiency virus-1 (HIV-1) samples to study its effects on HIV-1 sensitivity and to assay protein stability

Biochem/physiol Actions

Guanidine‐HCl (Gnd-HCl), an efficient chaotropic agent, is broadly used in RNA isolation, globular protein denaturation, and protein refolding studies as a denaturant. It is an appropriate sample buffer for the quick and effective elution of affinity-purified protein complexes and the subsequent protein digestion step.[1] GnHCl can be used to extract proteins in extracellular matrix (ECM)‐rich tissues including bone, cartilage, ligament, and tendon.[2] Gnd-HCl is not used in proteomics experiments due to its interference with trypsin activity at low molar concentrations and tendency to precipitate when combined with sodium dodecyl sulfate (SDS).[1] It is appropriate for performing rapid and single-step affinity-purification mass spectrometry (AP-MS) experiments.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianwen Xu et al.
Journal of pharmaceutical sciences, 110(12), 3819-3828 (2021-09-11)
The purpose of this investigation was to highlight the utility of nuclear magnetic resonance (NMR) as a multi-attribute method for the characterization of therapeutic antibodies. In this case study, we compared results from isothermal chemical denaturation (ICD) and NMR with
Nitish Agrawal et al.
PloS one, 6(6), e21339-e21339 (2011-07-09)
The HIV-1 envelope glycoprotein (Env) spike is challenging to study at the molecular level, due in part to its genetic variability, structural heterogeneity and lability. However, the extent of lability in Env function, particularly for primary isolates across clades, has
Jessica Colby-Milley et al.
PloS one, 10(6), e0130177-e0130177 (2015-06-16)
In addition to cognitive decline, individuals affected by Alzheimer's disease (AD) can experience important neuropsychiatric symptoms including sleep disturbances. We characterized the sleep-wake cycle in the TgCRND8 mouse model of AD, which overexpresses a mutant human form of amyloid precursor
Yalda Ashraf Kharaz et al.
Proteomics, 17(13-14) (2017-05-27)
Exploring the tendon proteome is a challenging but important task for understanding the mechanisms of physiological/pathological processes during ageing and disease and for the development of new treatments. Several extraction methods have been utilised for tendon mass spectrometry, however different
Jon W Poulsen et al.
Journal of proteome research, 12(2), 1020-1030 (2012-11-29)
Protein digestion is an integral part of the "shotgun" proteomics approach and commonly requires overnight incubation prior to mass spectrometry analysis. Quadruplicate "shotgun" proteomic analysis of whole yeast lysate demonstrated that Guanidine-Hydrochloride (Gnd-HCl) protein digestion can be optimally completed within

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service