Skip to Content
Merck
All Photos(1)

Key Documents

SYNF606150X

Synthware Buchner funnel

150 mL, porosity: extra coarse

Synonym(s):

Synthware F606150X, filtering funnel

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$106.00

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$106.00

About This Item

UNSPSC Code:
41121809
NACRES:
NB.51

$106.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

packaging

pkg of 1 ea

manufacturer/tradename

Synthware F606150X

capacity

150 mL
150 mL

Looking for similar products? Visit Product Comparison Guide

General description

With sealed-in glass disc of the listed porosity. Not recommended for filtration where pressure would exceed 15psi. XC-extra coarse, C-Coarse, M-Medium, F-Fine porosity.

Legal Information

Synthware is a trademark of Kemtech America Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service