Key Documents
Z553093
Sigma-Aldrich
Aldrich® Buchner funnel with joint and SafetyBarb® hose connector
capacity 60 mL, porosity 10-20 μm
Select a Size
1 EA
$206.00
$206.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$206.00
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
$206.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint ST/NS24/40
capacity
60 mL
pore size
10-20 μm porosity
General description
Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)