Key Documents
SYNA289250
Synthware anti-splash adapter
250 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 19/22
Synonym(s):
Synthware A289250, splash-guard adapter
Select a Size
1 EA
$129.00
$129.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$129.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$129.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
bottom joint: ST/NS19/22
joint: ST/NS top24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A289250
capacity
250 mL
Related Categories
General description
This design prevents possible contamination from rotary evaporator.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)