Key Documents
Z684511
Ace rotary evaporator bump trap
Select a Size
1 EA
$190.00
$190.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$190.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$190.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. What are the sizes of the tapered joint and the cone? These are not on the product description at time of asking.
    1 answer
    1. The product description of Z684511 is: Trap, bump, elliptical, 24/40 top, 24/40 bottom
      Helpful?