Key Documents
SYNA282914
Synthware anti-splash adapter
100 mL, top joint: ST/NS 29/42, bottom joint: ST/NS 14/20
Synonym(s):
Synthware A282914, anti-splash adapter, distillation adapter, adapter
Select a Size
1 EA
$271.00
$271.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$271.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$271.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
bottom joint: ST/NS14/20
joint: ST/NS top29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A282914
capacity
100 mL
Related Categories
General description
This design prevents possible contamination from rotary evaporator.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)