Key Documents
SYNA121420
Synthware vacuum transfer adapter with PTFE stopcocks
joint: ST/NS 14/20
Synonym(s):
Synthware A121420, glass vacuum transfer adapter
Select a Size
1 EA
$188.00
$188.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$188.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$188.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A121420
Related Categories
General description
With 2 mm PTFE stopcock. For transfer of sensitive and hazardous solutions.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)