Key Documents
SYNA682440
Synthware flushing adapter with PTFE stopcock
joint: ST/NS 24/40
Synonym(s):
Synthware A682440, adapter, flushing adapter
Select a Size
1 EA
$262.00
$262.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$262.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
$262.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A682440
General description
With 2 mm T-bore PTFE stopcock.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)