Skip to Content
Merck
All Photos(1)

Key Documents

M10204

Sigma-Aldrich

4-Methoxybenzenesulfonyl chloride

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$53.80
25 G
$110.00

$53.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$53.80
25 G
$110.00

About This Item

Linear Formula:
CH3OC6H4SO2Cl
CAS Number:
Molecular Weight:
206.65
Beilstein:
609005
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$53.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

bp

173 °C/14 mmHg (lit.)

mp

39-42 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)S(Cl)(=O)=O

InChI

1S/C7H7ClO3S/c1-11-6-2-4-7(5-3-6)12(8,9)10/h2-5H,1H3

InChI key

DTJVECUKADWGMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zafar Iqbal et al.
European journal of medicinal chemistry, 168, 154-175 (2019-03-01)
A novel class of spiroimidazolidine-2',4'-diones substituted with aryl sulfonyl group at different positions was designed and synthesized. The target compounds were evaluated for their potential to release insulin from MIN6 cell line derived from in-vivo immortalized insulin-secreting pancreatic cells. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service