Skip to Content
Merck
All Photos(2)

Documents

565849

Sigma-Aldrich

4-(Trifluoromethyl)benzenesulfonyl chloride

97%

Synonym(s):

α,α,α-Trifluoro-p-toluenesulfonyl chloride, 4-(Trifluoromethyl)benzene-1-sulfonyl chloride, [4-(Trifluoromethyl)phenyl]sulfonyl chloride, [p-(Trifluoromethyl)phenyl]sulfonyl chloride, p-Trifluoromethylbenzenesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4SO2Cl
CAS Number:
Molecular Weight:
244.62
Beilstein:
2113604
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

30-34 °C (lit.)

SMILES string

FC(F)(F)c1ccc(cc1)S(Cl)(=O)=O

InChI

1S/C7H4ClF3O2S/c8-14(12,13)6-3-1-5(2-4-6)7(9,10)11/h1-4H

InChI key

OZDCZHDOIBUGAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-(Trifluoromethyl)benzenesulfonyl chloride may be used to synthesize β-arylated thiophenes and 2,5-diarylated pyrrolevia palladium catalyzed desulfitative arylation of thiophenes and pyrroles, respectively.
4-(Trifluoromethyl)benzenesulfonyl chloride and N-vinylpyrrolidinone in acetonitrile can undergo photo-irradiation with visible light in the presence of Ir(ppy)2(dtbbpy)PF6 ([4,4′-bis(1,1-dimethylethyl)-2,2′-bipyridine-N1,N1′]bis[2-(2-pyridinyl-N)phenyl-C]iridium(III)hexafluorophosphate) and disodium phosphate to give the corresponding E-vinyl sulfone.

Caution

Low Melting Solid

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

"Benzenesulfonyl chlorides: new reagents for access to alternative regioisomers in palladium-catalysed direct arylations of thiophenes"
Yuan K, et al.
Chemical Science, 5(1), 392-396 (2014)
"C-H Functionalization of Enamides: Synthesis of ?-Amidovinyl Sulfones via Visible-Light Photoredox Catalysis"
Jiang H, et al.
Advanced Synthesis & Catalysis, 355(4), 809-813 (2013)
Palladium-Catalysed Direct Desulfitative Arylation of Pyrroles using Benzenesulfonyl Chlorides as Alternative Coupling Partners.
Jin R, et al.
Advanced Synthesis & Catalysis, 356(18), 3831-3841 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service