Skip to Content
Merck
All Photos(1)

Key Documents

L070000

Sigma-Aldrich

Fast Deprotection Cap A

45-55 g/L (TAC2O content, gravimetric)

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 L
$888.00
6 X 450 ML
$1,470.00

$888.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2.5 L
$888.00
6 X 450 ML
$1,470.00

About This Item

UNSPSC Code:
41116105
NACRES:
NA.21

$888.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

type

for DNA synthesis
for DNA synthesis

product line

Proligo Reagents

form

liquid

storage condition

dry at room temperature

concentration

45-55 g/L (TAC2O content, gravimetric)

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

Fast Deprotection Cap A contains tert-butylphenoxyacetyl aceticanhydride (TAC2O) in tetrahydrofuran, which ensures thedisplacement of tert-butylphenoxyacetyl (TAC) on guanine bases doesnot occur.

Preparation Note

Tetrahydrofurane / TAC2O 100/5 (v/w)

Other Notes

Fast Deprotection Cap A solution must be used for synthesis with Sigma-Aldrich′s TAC-protected phosphoramidites. Fast Deprotection Cap A solution, containing tert-butylphenoxyacetyl acetic anhydride (TAC2O) in tetrahydrofuran, is used in place of Cap A solution to ensure that the displacement of tert-butylphenoxyacetyl (TAC) on guanine bases of the TAC-protected DNA and RNA phosphoramidites does not occur.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

6.1 °F

Flash Point(C)

-14.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service