Skip to Content
Merck
All Photos(1)

Key Documents

L070030

Sigma-Aldrich

Fast Deprotection Cap A

configured for ABI

Sign Into View Organizational & Contract Pricing

Select a Size

6 X 180 ML
$590.00

$590.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
6 X 180 ML
$590.00

About This Item

UNSPSC Code:
41116105
NACRES:
NA.21

$590.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

type

for DNA synthesis
for DNA synthesis

product line

Proligo Reagents

storage condition

dry at room temperature

concentration

45-55 g/L (TAC2O content, gravimetric)

color

clear

compatibility

configured for ABI

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Tetrahydrofurane / TAC2O 100/5 (v/w)

Other Notes

Fast Deprotection Cap A solution must be used for synthesis with Sigma-Aldrich′s TAC-protected phosphoramidites. Fast Deprotection Cap A solution, containing tert-butylphenoxyacetyl acetic anhydride (TAC2O) in tetrahydrofuran, is used in place of Cap A solution to ensure that the displacement of tert-butylphenoxyacetyl (TAC) on guanine bases of the TAC-protected DNA and RNA phosphoramidites does not occur.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

6.1 °F

Flash Point(C)

-14.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service