Skip to Content
Merck
All Photos(1)

Key Documents

JKA3033

Sigma-Aldrich

mPEG10K

average Mn 10,000, propionaldehyde, amine reactive

Synonym(s):

mPEG-Propionaldehyde, Polyethylene glycol, mPEG-ALD, Methoxy-PEG-Propionaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$689.00

$689.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$689.00

About This Item

Linear Formula:
CH3O(CH2CH2O)nCH2CH2COH
UNSPSC Code:
12162002
NACRES:
NA.23

$689.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

mPEG10K-Propionaldehyde, average Mn 10,000

form

solid

mol wt

average Mn 10,000

reaction suitability

reagent type: chemical modification reagent
reactivity: N-terminal amine reactive

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Methoxy PEG Propionaldehyde. N-terminal amine reactive PEG; reacts at pH 5-8 in the presence of a reducing reagent.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue, X., et al.
Process. Biochem., 49(7) (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service