Skip to Content
Merck
All Photos(1)

Key Documents

JKA3060

Sigma-Aldrich

mPEG5K

average Mn 5,000, Hydrazide, carboxyl reactive

Synonym(s):

mPEG-Hydrazide, Methoxy-PEG-HZ, Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$700.00

$700.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$700.00

About This Item

Linear Formula:
CH3O(CH2CH2O)nCH2CONHNH2
UNSPSC Code:
12162002
NACRES:
NA.23

$700.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

mPEG5K-Hydrazide, average Mn 5,000

form

solid

mol wt

average Mn 5,000

reaction suitability

reagent type: chemical modification reagent
reactivity: carboxyl reactive

polymer architecture

shape: linear
functionality: monofunctional

storage temp.

−20°C

General description

Methoxy PEG Hydrazide reacts with aldehydes and ketones to give stable hydrazones in a single step; it can also react with activated carboxylic acids.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhou, Z., et al.
Bioconjugate Chemistry, 25(1) (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service