CLS4517
Corning® High Content Imaging Microplate
black cyclic olefin copolymer (COC), flat bottom, sterile, film bottom, lid, pack of 20
Synonym(s):
96 multiwell plate
Select a Size
$778.00
Select a Size
About This Item
clear bottom
flat bottom
flat bottom wells
$778.00
Recommended Products
material
black cyclic olefin copolymer (COC)
clear bottom
flat bottom
flat bottom wells
sterility
sterile
feature
lid
film bottom
low base
packaging
pack of 20 plates
pack of 20
manufacturer/tradename
Corning 4517
surface area
0.16 cm2 , cell growth area
wall volume
190 μL
wall working volume
50 μL
suitability
suitable for (cell cuture)
Related Categories
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Customers Also Viewed
Articles
A frequently asked questions guide for users of the Corning® high content screening microplates.