Skip to Content
Merck
All Photos(1)

Key Documents

I22900

Sigma-Aldrich

1,2-O-Isopropylidene-α-D-glucofuranose

98%

Synonym(s):

Monoacetone glucose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O6
CAS Number:
Molecular Weight:
220.22
Beilstein:
82670
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D −13°, c = 1 in H2O

mp

159-160 °C (lit.)

SMILES string

CC1(C)O[C@H]2O[C@H]([C@H](O)CO)[C@H](O)[C@H]2O1

InChI

1S/C9H16O6/c1-9(2)14-7-5(12)6(4(11)3-10)13-8(7)15-9/h4-8,10-12H,3H2,1-2H3/t4-,5+,6-,7-,8-/m1/s1

InChI key

BGGCXQKYCBBHAH-OZRXBMAMSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S H Cmelik
Zentralblatt fur Bakteriologie. 1. Abt. Originale. A: Medizinische Mikrobiologie, Infektionskrankheiten und Parasitologie, 247(4), 495-501 (1980-01-01)
Various species of Clostridium, Bacteroides, Propionibacterium and Eubacterium were incubated in a 1% solution of 1,2-O-iso-propylidene-D-glucofuranose in a peptone-yeast-extract (PY) medium according to the VPI-technique. The volatile and non volatile acids were investigated by gas-liquid chromatography. All microorganisms showed a
Aifric O'Sullivan et al.
Molecular nutrition & food research, 55(7), 1018-1025 (2011-04-27)
Research reports suggest that vitamin D affects glucose and insulin metabolism; however, the exact mechanisms are unclear. ²H NMR analysis of monoacetone glucose (MAG) after tracer administration provides a non-invasive method of profiling hepatic glucose metabolism. This study examined the
G N Richards et al.
Carbohydrate research, 219, 91-99 (1991-10-14)
Methyl alpha,beta-D-glucofuranoside (1) and 1,2-O-isopropylidene-alpha-D-glucofuranose (2) have been polymerized thermally, using 0.1% phosphoric acid as catalyst. The resulting alcohol-insoluble polymers were analyzed by gel-permeation chromatography and methylation analysis. The two starting materials gave polymers that were very similar in molecular-weight
Tiago Cardoso Alves et al.
NMR in biomedicine, 21(4), 337-344 (2007-08-09)
The gluconeogenic contribution to glucose production in livers isolated from rats fasted for 24 h was determined by 13C-NMR isotopomer distribution analysis of secreted glucose enriched from 99% [13C]bicarbonate (n = 4) and 99% [1-13C]lactate (n = 4). Experiments with
Takashi Kobayashi et al.
Biotechnology letters, 32(11), 1679-1684 (2010-06-25)
In order to synthesize a sugar ester at high concentration, 1,2-O-isopropylidene-α-D-glucofuranose (IpGlc), which is one of the sugar acetals and is more hydrophobic than unmodified glucose, was esterified with palmitic acid at 40°C using immobilized lipase from Candida antarctica in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service