Skip to Content
Merck
All Photos(1)

Documents

H9021

Sigma-Aldrich

Methyl cis-10-heptadecenoate

≥99% (GC), liquid

Synonym(s):

cis-10-Heptadecenoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H34O2
CAS Number:
Molecular Weight:
282.46
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99% (GC)

form

liquid

storage temp.

−20°C

SMILES string

CCCCCC\C=C/CCCCCCCCC(=O)OC

InChI

1S/C18H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-2/h8-9H,3-7,10-17H2,1-2H3/b9-8-

InChI key

JNSUZRHLHDQGPN-HJWRWDBZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin N Andersson et al.
The Journal of experimental biology, 221(Pt 24) (2018-10-27)
Diet and ambient temperature affect animal physiology, survival and reproductive success. However, knowledge of how these environmental factors interact to shape physiological processes and life-history traits of birds and other animals is largely lacking. By exposing adult great tits (Parus
Cas Eikenaar et al.
Physiological and biochemical zoology : PBZ, 90(2), 223-229 (2017-03-10)
Most avian migrants alternate flight bouts, characterized by high metabolic rates, with stopovers, periods of fuel replenishment through hyperphagia. High-energy metabolism and excessive calorie intake shift the balance between damaging prooxidants and antioxidants toward the former. Hence, migration likely affects
Edward Mubiru et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(6), 1000-1011 (2017-03-30)
Epoxy fatty acids (EFAs) are secondary oxidation products formed from unsaturated fatty acid hydroperoxides. Seventeen food categories were analysed for C18 monoEFAs of food products available on the Belgian market. A quantitative exposure assessment was performed based on deterministic and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service