Skip to Content
Merck
All Photos(4)

Key Documents

M3378

Sigma-Aldrich

Methyl myristate

≥99% (GC)

Synonym(s):

Methyl tetradecanoate, Myristic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$77.80
25 G
$126.00

$77.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$77.80
25 G
$126.00

About This Item

Linear Formula:
CH3(CH2)12COOCH3
CAS Number:
Molecular Weight:
242.40
Beilstein:
1773739
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

$77.80

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

biological source

synthetic (organic)

Quality Level

Assay

≥99% (GC)

form

liquid

refractive index

n20/D 1.436 (lit.)

bp

323 °C (lit.)

mp

18 °C (lit.)

density

0.855 g/mL at 25 °C (lit.)

functional group

ester

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCC(=O)OC

InChI

1S/C15H30O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17-2/h3-14H2,1-2H3

InChI key

ZAZKJZBWRNNLDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl myristate may be used as an intermediate polarity oil component of oil-in-water, especially “Pickering-type”, emulsions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Walsh et al.
Langmuir : the ACS journal of surfaces and colloids, 26(23), 18039-18048 (2010-11-11)
Sterically stabilized polystyrene latexes were prepared by aqueous emulsion polymerization using a poly(ethylene imine) (PEI) stabilizer in the presence of 4-vinylbenzyl chloride (4-VBC; 1.0 wt % based on styrene). Partial quaternization of the amine groups on the PEI chains by
Tomohiro Imura et al.
Journal of oleo science, 57(8), 415-422 (2008-07-16)
Mannosylerythritol lipid-A (MEL-A) is one of the most promising glycolipid biosurfactants, and abundantly produced by Pseudozyma yeasts. MEL-A gives not only excellent self-assembling properties but also a high binding affinity toward human immunoglobulin G (HIgG). In this study, three kinds
E S Read et al.
Langmuir : the ACS journal of surfaces and colloids, 20(18), 7422-7429 (2004-08-25)
Sterically stabilized polystyrene latexes (previously described by Amalvy, J. I.; et al. Chem. Commun. 2003, 1826) were evaluated as pH-responsive particulate emulsifiers for the preparation of both oil-in-water and water-in-oil emulsions. The steric stabilizer was a well-defined AB diblock copolymer
Bernard P Binks et al.
Langmuir : the ACS journal of surfaces and colloids, 22(5), 2050-2057 (2006-02-24)
Aqueous dispersions of lightly cross-linked poly(4-vinylpyridine)/silica nanocomposite microgel particles are used as a sole emulsifier of methyl myristate and water (1:1 by volume) at various pH values and salt concentrations at 20 degrees C. These particles become swollen at low
Syuji Fujii et al.
Journal of colloid and interface science, 315(1), 287-296 (2007-08-08)
Hydroxyapatite (HAp) nanoparticles with spherical, rod-shaped or fiber-shaped morphologies were synthesized by wet chemical method in aqueous media. Scanning electron microscopy, dynamic light scattering, helium pycnometry, and aqueous electrophoresis techniques were used to characterize the nanoparticles in terms of their

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service