Skip to Content
Merck

E2012

Methyl all-cis-5,8,11,14,17-eicosapentaenoate

≥97% (capillary GC)

Synonym(s):

cis-5,8,11,14,17-Eicosapentaenoic acid methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
CH3(CH2CH=CH)5(CH2)3CO2CH3
CAS Number:
Molecular Weight:
316.48
UNSPSC Code:
12352211
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
1914828

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

assay

≥97% (capillary GC)

form

oil

functional group

ester

lipid type

omega FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)OC

InChI

1S/C21H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(22)23-2/h4-5,7-8,10-11,13-14,16-17H,3,6,9,12,15,18-20H2,1-2H3/b5-4-,8-7-,11-10-,14-13-,17-16-

InChI key

QWDCYFDDFPWISL-JEBPEJKESA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
P9667R8750E6885
assay

≥97% (capillary GC)

assay

≥99% (capillary GC)

assay

≥99% (GC)

assay

≥98%

functional group

ester

functional group

ester

functional group

ester

functional group

ester

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

form

oil

form

liquid

form

liquid

form

liquid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

Packaging

Sealed ampule under argon.

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

219.2 °F - closed cup

flash_point_c

104.0 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service