Skip to Content
Merck
All Photos(4)

Key Documents

143758

Sigma-Aldrich

3,4-Dimethoxybenzaldehyde

99%

Synonym(s):

Methylvanillin, NSC 24521, NSC 8500, Vanillin methyl ether, Veratraldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$75.30
100 G
$87.30
500 G
$137.00

$75.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$75.30
100 G
$87.30
500 G
$137.00

About This Item

Linear Formula:
(CH3O)2C6H3CHO
CAS Number:
Molecular Weight:
166.17
Beilstein:
473899
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$75.30

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

bp

281 °C (lit.)

mp

40-43 °C (lit.)

solubility

alcohol: freely soluble
diethyl ether: freely soluble
water: slightly soluble (hot)

functional group

aldehyde

SMILES string

[H]C(=O)c1ccc(OC)c(OC)c1

InChI

1S/C9H10O3/c1-11-8-4-3-7(6-10)5-9(8)12-2/h3-6H,1-2H3

InChI key

WJUFSDZVCOTFON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Dimethoxybenzaldehyde forms 1:1 inclusion complexes with cyclodextrins[1]. It reacts with 3-acetyl-2,5-dimethythiophene to yield chalcone dye, (2E)-3-(3,4-Dimethoxyphenyl)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one[2].

Application

3,4-Dimethoxybenzaldehyde was used in the preparation of 4-chloromethyl-2-(dimethoxyphenyl)-1,3-dioxolane[3]. It was used in the synthesis of (+)-lithospermic acid, having anti-HIV activity[4].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2-(Multimethoxy) phenyl-4-methylene-1, 3-dioxolane (I): Preparation and Cationic Polymerization of 2-(Dimethoxy) phenyl-4-MDO Derivatives.
Kim JT and Gong M-S.
Bull. Korean Chem. Soc., 20, 663-666 (1999)
Tirumala G Varadaraju et al.
Organic & biomolecular chemistry, 10(28), 5456-5465 (2012-06-07)
An efficient and convergent route for the synthesis of the natural product (+)-lithospermic acid, which possesses anti-HIV activity, was accomplished. The (±)-trans-dihydrobenzo[b]furan core therein was prepared by two different strategies. The first strategy involved the use of a palladium-catalyzed annulation
M Jude Jenita et al.
Journal of fluorescence, 24(3), 695-707 (2013-12-07)
The inclusion complexation of 2-hydroxy-3-methoxybenzaldehyde (2HMB), 4-hydroxy-3-methoxybenzaldehyde (4HMB), 3,4-dimethoxybenzaldehyde (DMB) and 4-hydroxy-3,5-dimethoxybenzaldehyde (HDMB) with α-CD, β-CD, HP-α-CD and HP-β-CD were carried out by UV-Visible, steady-state and time-resolved fluorescence and PM3 methods. All the benzaldehydes shows dual fluorescence in aqueous and
Abdullah M Asiri et al.
Journal of fluorescence, 23(6), 1271-1278 (2013-07-13)
This study introduced spectroscopic properties, physicochemical parameters, and polarity and photostability behaviors of a newly prepared chalcone dye. The chalcone dye, (2E)-3-(3,4-Dimethoxyphenyl)-1-(2,5-dimethylthiophen-3-yl)prop-2-en-1-one (DDTP), was synthesized by the reaction of 3,4-dimethoxybenzaldehyde with 3-acetyl-2,5-dimethythiophene. Results of FT-IR, (1)H-NMR, (13)C-NMR and elemental analysis
Francesca D'Acunzo et al.
Biochemical and biophysical research communications, 313(1), 17-21 (2003-12-16)
Veratrylchitosan, a polysaccharide-supported lignin model compound, has been synthesised by covalently attaching 3-(3,4-dimethoxybenzyloxy)propionic acid to the polysaccharide chitosan through an amide linkage. When this polymer was used as a substrate in the oxidation promoted by lignin peroxidase (LiP), significant decomposition

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service