Skip to Content
Merck
All Photos(2)

Key Documents

F4807

Sigma-Aldrich

2-Fluorobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$38.00
100 G
$91.80

$38.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$38.00
100 G
$91.80

About This Item

Linear Formula:
FC6H4CHO
CAS Number:
Molecular Weight:
124.11
Beilstein:
507155
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$38.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.521 (lit.)

bp

90-91 °C/46 mmHg (lit.)

mp

−44.5 °C (lit.)

density

1.178 g/mL at 25 °C (lit.)

SMILES string

[H]C(=O)c1ccccc1F

InChI

1S/C7H5FO/c8-7-4-2-1-3-6(7)5-9/h1-5H

InChI key

ZWDVQMVZZYIAHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiafeng Ding et al.
Environmental science and pollution research international, 25(30), 30160-30169 (2018-08-29)
Hydroxylated polybrominated diphenyl ethers (OH-PBDEs) are ubiquitous and highly toxic emerging endocrine disruptors found in surface and subsurface soils and clay deposits. Seriously, they could be easily transformed to the more toxic dioxins (PBDD/Fs) in photochemical processes and incineration, but
Yuan Chen et al.
Polymers, 11(6) (2019-06-07)
In this paper, we report chemical modifications of inulin by seven kinds of aromatic Schiff bases, which are different from their substituent groups. The obtained inulin derivatives were confirmed by FTIR, 1H NMR, and 13C NMR. Then, we studied their

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service