Skip to Content
Merck
All Photos(1)

Key Documents

290173

Sigma-Aldrich

3,5-Difluorobenzaldehyde

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$71.40

$71.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$71.40

About This Item

Linear Formula:
F2C6H3CHO
CAS Number:
Molecular Weight:
142.10
Beilstein:
2573392
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$71.40

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

refractive index

n20/D 1.493 (lit.)

density

1.296 g/mL at 20 °C (lit.)

functional group

aldehyde
fluoro

SMILES string

[H]C(=O)c1cc(F)cc(F)c1

InChI

1S/C7H4F2O/c8-6-1-5(4-10)2-7(9)3-6/h1-4H

InChI key

ASOFZHSTJHGQDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The 19F{′H} spectra of 3,5-difluorobenzaldehyde at low temperature in dimethyl ether solution was studied[1].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A positive 6 J (H, CHO) in some meta derivatives of benzaldehyde. A simple model.
Canadian Journal of Chemistry, 67(5), 827-830 (1989)
Rasa Keruckiene et al.
Beilstein journal of organic chemistry, 16, 1142-1153 (2020-06-20)
Three compounds, bearing a quinazoline unit as the acceptor core and carbazole, dimethyldihydroacridine, or phenothiazine donor moieties, were designed and synthesized in two steps including a facile copper-catalyzed cyclization and a nucleophilic aromatic substitution reaction. The photophysical properties of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service