Skip to Content
Merck
All Photos(2)

Key Documents

D9272

Sigma-Aldrich

N-(2,6-Diisopropylphenylcarbamoylmethyl)iminodiacetic acid

≥98%

Synonym(s):

N-[2-[[2,6-Bis(1-methylethyl)phenyl]amino]-2-oxoethyl]-N-(carboxymethyl)glycine, DISIDA

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$217.00

$217.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$217.00

About This Item

Empirical Formula (Hill Notation):
C18H26N2O5
CAS Number:
Molecular Weight:
350.41
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$217.00


Estimated to ship onApril 16, 2025


Request a Bulk Order

Quality Level

Assay

≥98%

form

powder

SMILES string

CC(C)c1cccc(C(C)C)c1NC(=O)CN(CC(O)=O)CC(O)=O

InChI

1S/C18H26N2O5/c1-11(2)13-6-5-7-14(12(3)4)18(13)19-15(21)8-20(9-16(22)23)10-17(24)25/h5-7,11-12H,8-10H2,1-4H3,(H,19,21)(H,22,23)(H,24,25)

InChI key

UDUSOMRJOPCWHT-UHFFFAOYSA-N

Application

Involved in Pharmacological studies as a ligand for identifying molecular structures for advances melanoma

Reactant involved in:
  • Synthesis of N-(2,6-diisopropylacetanilido)-iminodiacetic acid derivatives and labeling for studies of possible hepatobiliary imaging agents
  • Preparing labeled radiopharmaceuticals using a borohydride exchange resin
  • Evaluation of radiopharmaceutical for cholescintigraphy

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service