Skip to Content
Merck
All Photos(1)

Key Documents

224057

Sigma-Aldrich

1-Chloro-4-fluorobenzene

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$76.70

$76.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$76.70

About This Item

Linear Formula:
ClC6H4F
CAS Number:
Molecular Weight:
130.55
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$76.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.495 (lit.)

bp

129-130 °C (lit.)

mp

−27-−26 °C (lit.)

density

1.226 g/mL at 25 °C (lit.)

functional group

chloro
fluoro

SMILES string

Fc1ccc(Cl)cc1

InChI

1S/C6H4ClF/c7-5-1-3-6(8)4-2-5/h1-4H

InChI key

RJCGZNCCVKIBHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David J Kemp et al.
Physical chemistry chemical physics : PCCP, 20(18), 12503-12516 (2018-05-01)
The vibrations of the ground state cation (X[combining tilde]2B1) of para-chlorofluorobenzene (pClFB) have been investigated using zero-electron-kinetic-energy (ZEKE) spectroscopy. ZEKE spectra were recorded using different vibrational levels of the S1 state as intermediate levels, for which assignments were put forward
Asha Lal et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 24(2), 231-236 (2008-02-14)
A new extraction and cleanup procedure with gas chromatography was developed for the sensitive determination of acephate, dimethoate, malathion, diazinon, quinalphos, chlorpyrifos, profenofos, alpha-endosulfan, beta-endosulfan, chlorothalonil and carbaryl using 1-chloro-4-fluorobenzene as an internal standard in fruits and vegetables. Several extracting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service