CLS316610
Pyrex® cloning cylinder
O.D. × H 10 mm × 10 mm
Synonym(s):
lab equipment accessory
Select a Size
$262.00
Select a Size
About This Item
$262.00
Recommended Products
material
borosilicate glass
cylindrical
sterility
non-sterile
packaging
pack of 125
manufacturer/tradename
Corning 3166-10
O.D. × H
10 mm × 10 mm
General description
Features and Benefits
- Pyrex® cloning cylinder Type I Class A - Low-Expansion Borosilicate Glass
- Compatible with heating using hot plate, flame, oven, microwave, or heating mantles.
- Product can be cooled to -230°C.
- Product Dimensions - 10 mm OD x 10 mm High.
- DO NOT plunge product directly into a liquid coolant.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)