Skip to Content
Merck
All Photos(8)

Key Documents

B4252

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside

5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside
1 of 1 reviewers received a sample product or took part in a promotion

chromogenic, ≥98%, powder

Synonym(s):

5-Bromo-4-chloro-3-indolyl β-D-galactoside, BCIG, X-Gal

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$117.00
50 MG
$186.00
250 MG
$335.00
500 MG
$697.00
1 G
$1,060.00
2 G
$1,910.00
5 G
$4,190.00

$117.00


In StockDetails



Select a Size

Change View
100 MG
$117.00
50 MG
$186.00
250 MG
$335.00
500 MG
$697.00
1 G
$1,060.00
2 G
$1,910.00
5 G
$4,190.00

About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
Beilstein:
1402009
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

$117.00


In StockDetails


Product Name

5-Bromo-4-chloro-3-indolyl β-D-galactopyranoside, ≥98%, powder

Quality Level

Assay

≥98%

form

powder

solubility

DMF: methanol (1:1): 50 mg/mL

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2c[nH]c3ccc(Br)c(Cl)c23)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C14H15BrClNO6/c15-5-1-2-6-9(10(5)16)7(3-17-6)22-14-13(21)12(20)11(19)8(4-18)23-14/h1-3,8,11-14,17-21H,4H2/t8-,11+,12+,13-,14-/m1/s1

InChI key

OPIFSICVWOWJMJ-AEOCFKNESA-N

Looking for similar products? Visit Product Comparison Guide

Application

X-Gal is a chromogenic substrate for β-galactosidase that produces a rich blue color that can easily be detected visually over background. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana Klein et al.
Antioxidants & redox signaling, 24(2), 53-69 (2015-06-13)
Previous thorax irradiation promotes metastatic spread of tumor cells to the lung. We hypothesized that vascular damage facilitates lung metastasis after thorax irradiation and that therapeutically applied multipotent mesenchymal stromal cells (MSCs) with reported repair activity may prevent these adverse
Lía Alza et al.
Cancers, 12(5) (2020-05-01)
Focal adhesion kinase (FAK) is a central component of focal adhesions that regulate cancer cell proliferation and migration. Here, we studied the effects of FAK inhibition in glioblastoma (GBM), a fast growing brain tumor that has a poor prognosis. Treating
Carina Henning et al.
Scientific reports, 9(1), 16931-16931 (2019-11-16)
An acute increase in blood flow triggers flow-mediated dilation (FMD), which is mainly mediated by endothelial nitric oxide synthase (eNOS). A long-term increase in blood flow chronically enlarges the arterial lumen, a process called arteriogenesis. In several common human diseases
Elijah D Lowenstein et al.
eLife, 10 (2021-02-17)
The mature cerebellum controls motor skill precision and participates in other sophisticated brain functions that include learning, cognition, and speech. Different types of GABAergic and glutamatergic cerebellar neurons originate in temporal order from two progenitor niches, the ventricular zone and
Enzo R Porrello et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(1), 187-192 (2012-12-19)
We recently identified a brief time period during postnatal development when the mammalian heart retains significant regenerative potential after amputation of the ventricular apex. However, one major unresolved question is whether the neonatal mouse heart can also regenerate in response

Questions

  1. Could you Please provide information on the solvent used for item B4252-50MG ?

    1 answer
    1. This compound is soluble in Dimethylformamide:methanol (1:1) at 50 mg/mL, Dimethylformamide at 20 mg/mL, and DMSO at 20 mg/mL. When protected from light, stock solutions of X-gal in the given solvents are stable for up to 2 months at 2 to 8 °C and up to 1 year in aliquots at -20 °C.

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Spain
    • Review 1
    • Votes 0
    5 out of 5 stars.

    X-Gal

    We used to stain tissue samples from mice and it worked perfectly well!

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service