Skip to Content
Merck
All Photos(1)

Key Documents

C7983

Sigma-Aldrich

Scienceware® cloning cylinders, polystyrene

I.D. × H 4.7 mm × 8 mm

Synonym(s):

cloning cylinder, cloning equipment

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
$110.00

$110.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
50 EA
$110.00

About This Item

UNSPSC Code:
41106618
NACRES:
NB.22

$110.00


Estimated to ship onMay 29, 2025


sterility

sterile

packaging

pack of 50 ea

manufacturer/tradename

Bel-Art F37847-0100

I.D. × H

4.7 mm × 8 mm

Looking for similar products? Visit Product Comparison Guide

General description

To isolate a clone, sterile grease (not included) is applied to bottom edge of cylinder and inverted over clone of choice. Provides an isolation chamber to trypsinize and recover clonal colonies.

Legal Information

Scienceware is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of colony-forming cells in adult human articular cartilage.
Ozbey O, Sahin Z, Acar N, et al.
Acta Histochemica, 116(5), 763-770 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service