Skip to Content
Merck
All Photos(1)

Key Documents

C25201

Sigma-Aldrich

5-Chlorobenzotriazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClN3
CAS Number:
Molecular Weight:
153.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

157-159 °C (lit.)

SMILES string

Clc1ccc2[nH]nnc2c1

InChI

1S/C6H4ClN3/c7-4-1-2-5-6(3-4)9-10-8-5/h1-3H,(H,8,9,10)

InChI key

PZBQVZFITSVHAW-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chien-Hao Chen et al.
Food chemistry, 333, 127516-127516 (2020-07-20)
Benzotriazoles (BTRs) and benzothiazoles (BTs) are two groups of emerging concern and high production volume contaminants. Via the biomagnification of the food web, they could jeopardize human health. In this work, rapid determining the presence of five BTRs and two
Peter J E M van der Linden et al.
Lab on a chip, 20(22), 4128-4140 (2020-10-16)
The use of microfluidics on synchrotron X-ray beamlines represents an advanced sample preparation and delivery platform for state-of-the-art X-ray characterization of micro-samples. The recent developments of 3D printing technologies have opened possibilities for rapid fabrication of complex microfluidic devices. One
Jiufeng Li et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1070, 70-75 (2017-11-02)
Benzotriazole (BTR) and benzothiazole (BTH) derivatives are extensively applied in industrial processes and consumer products, and are thus frequently detected in the environmental matrices. Due to their potential estrogenic effects reported in animal studies, the assessment of human exposure to
Manuel Nieto-Domínguez et al.
Nature communications, 11(1), 4864-4864 (2020-09-27)
The synthesis of customized glycoconjugates constitutes a major goal for biocatalysis. To this end, engineered glycosidases have received great attention and, among them, thioglycoligases have proved useful to connect carbohydrates to non-sugar acceptors. However, hitherto the scope of these biocatalysts
Attilio Naccarato et al.
Journal of chromatography. A, 1338, 164-173 (2014-03-19)
This work proposes a new approach for the simultaneous determination of benzothiazoles, benzotriazoles and benzosulfonamides in different environmental matrices and human urine, using solid-phase microextraction coupled with gas-chromatography-triple quadrupole mass spectrometry (SPME-GC-QqQMS). The analytes object of this investigation have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service