Recommended Products
Assay
99%
form
solid
SMILES string
OC(=O)c1ccc2[nH]nnc2c1
InChI
1S/C7H5N3O2/c11-7(12)4-1-2-5-6(3-4)9-10-8-5/h1-3H,(H,11,12)(H,8,9,10)
InChI key
GUOVBFFLXKJFEE-UHFFFAOYSA-N
Gene Information
human ... GPR109B(8843)
Related Categories
Application
Benzotriazole-5-carboxylic acid was used as bifunctional ligand in solvent induced synthesis of diverse dimensional coordination assemblies of cupric benzotriazole-5-carboxylate. It was also used in hydrothermal synthesis of new coordination polymer, [Zn(Btca)(Phen)]n (H2Btca = benzotriazole-5-carboxylic acid, Phen = 1,10-phenanthroline).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of medicinal chemistry, 56(5), 1865-1877 (2013-02-09)
We have reported that retinoid X receptor (RXR) partial agonist 1-(3,5,5,8,8-pentamethyl-5,6,7,8-tetrahydro-2-naphthyl)-1H-benzotriazole-5-carboxylic acid (CBt-PMN, 4a) shows a significant antidiabetes effect in the KK-A(y) type 2 diabetes model mice, with reduced side effects compared to RXR full agonists. To elucidate the mechanism
Inorganic chemistry, 50(21), 11032-11038 (2011-10-11)
Three cupric coordination assemblies [Cu(btca)(H(2)O)(2)] (1), [Cu(btca)(H(2)O)(3.5)](8)·16H(2)O (2), and [Cu(2.5)(btca)(1.5)(Hbtca)(0.5)(μ-Cl)(0.5)(μ(3)-OH)(H(2)O)]·H(2)O (3) have been solvothermally synthesized by cupric salts and a bifunctional ligand benzotriazole-5-carboxylic acid (H(2)btca) in different solvent medium. These complexes were structurally characterized by X-ray diffraction analyses and further
Hydrothermal synthesis and crystal structure of a new 2D metal-organic framework:[Zn (Btca)(Phen)] n (H2Btca= benzotriazole-5-carboxylic acid, phen= 1, 10-phenanthroline).
Russian Journal of Coordination Chemistry, 37(3), 172-175 (2011)
Archiv der Pharmazie, 322(8), 457-459 (1989-08-01)
The 1H-benzotriazolecarboxylic acids 1 and 2 were stable towards oxidative metabolism as well in vitro as in vivo. In the in vivo studies 2.25% of the ester glucuronide of 1H-benzotriazole-5-carboxylic acid was found in urine.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service