Skip to Content
Merck
All Photos(1)

Key Documents

BR781614

BRAND® microplate BRANDplates®, pureGrade

size 96 wells, size 350 μL, foil bottom, transparent

Synonym(s):

BRANDplates®, Microplates, medium binding, pureGrade

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
$1,160.00

$1,160.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
50 EA
$1,160.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
material:
colorless wells
foil bottom
polystyrene
size:
350 μL
96 wells

$1,160.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

colorless wells
foil bottom
polystyrene

sterility

non-sterile

feature

lid: no
skirt Skirt (F-bottom)
UV transparent plates

packaging

pack of 100 ea

manufacturer/tradename

BRAND 781614

size

350 μL
96 wells

working volume

350 μL

color

transparent

suitability

suitable for (bacteriological assays)

binding type

medium binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Clearly distinguishable via color code: grey embossed alphanumeric coding for 96-well standard plates in white or black

Features and Benefits

  • Optimal surfaces for optimal results
  • Reliable quality from the cleanroom
  • Tested by independent laboratories

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service