Skip to Content
Merck
All Photos(1)

Key Documents

BR781602

BRAND® microplate BRANDplates®, pureGrade

size 96 wells, size 350 μL, flat bottom, transparent

Synonym(s):

96-well, BRANDplates®, Microplates, Plates, medium binding, pureGrade

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$211.00

$211.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025



Select a Size

Change View
100 EA
$211.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
colorless wells
flat bottom
polystyrene
size:
350 μL
96 wells

$211.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


material

colorless wells
flat bottom
polystyrene

sterility

non-sterile

feature

lid: no
skirt Skirt (F-bottom)

packaging

pack of 100 ea

manufacturer/tradename

BRAND 781602

size

350 μL
96 wells

working volume

350 μL

color

transparent

suitability

suitable for (bacteriological assays)

binding type

medium binding surface

Looking for similar products? Visit Product Comparison Guide

General description

The PS 96-well multiwell plates, branded as BIO-CERT® CERTIFIED QUALITY, offer a high-quality solution for a variety of laboratory applications. With their exceptional purity and quality assurance, these multiwell plates are an indispensable tool for researchers and scientists working in pharmaceutical, biotechnological, and life science fields.

Application

Spectroscopy assays, assays, titrations, sample collection, sample storage

Features and Benefits

  • Optimal surfaces for optimal results
  • Tested by independent laboratories
  • BRANDplates® pureGrade microtitration plates are manufactured from high purity, crystal-clear polystyrene.
  • Cleanroom production ensures the best possible cleanliness.
  • Medium binding, non-treated, non-sterile surface
  • BIO-CERT® CERTIFIED QUALITY: Free of human DNA, RNase, DNase and Pyrogens
  • Best suited for homogenous assays, screening, and storage.

Legal Information

BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service