Skip to Content
Merck
All Photos(1)

Key Documents

BR781332

BRAND® PCR tubes, strips of 8

attached single caps, standard profile, capacity 0.2 mL

Synonym(s):

BIO-CERT, PCR, PCR Tubes, Tube Strips, Tubes

Sign Into View Organizational & Contract Pricing

Select a Size

120 EA
$537.00

$537.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
120 EA
$537.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

$537.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

material

cap (Attached single caps)
clear
polypropylene tube

description

DNase-/RNase- DNA-free

sterility

non-sterile

feature

neck thread push fit
attached single caps

packaging

pack of 120

manufacturer/tradename

BRAND 781332

technique(s)

PCR: suitable

capacity

0.2 mL

working volume

0.2 mL

suitability

suitable for (for RealTime-PCR)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

PCR 8- and 12-tube strips offer a flexible solution for your PCR or qPCR. Domed caps provide extra sealing pressure via the lid of the thermal cycler. Flat caps are required for qPCR. Attached cap strips can be easily opened and closed with one hand. Separate cap and film strips are suitable for automation, as they won′t obstruct robotics. No matter which cap you choose, all variants seal the vessels reliably, effectively protecting your samples from evaporation and contamination. The extra-thin walls allow for even heat transfer during the process of heating and cooling for short cycle time.

Application

PCR, qPCR (flat caps only)

Features and Benefits

  • High flexibility with a wide range of cap types and film strips
  • Reduced evaporation losses thanks to tightly closing cap strips
  • Rapid heat transfer through thin vessel walls
  • Numbered wells for clear identification
  • Free of: DNA, RNA, DNase, RNase, pyrogens, PCR inhibitors

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service