BR129055
BRAND® bottle for Schilling burettes
volume 500 mL, low-density polyethylene
Synonym(s):
Accessories, Bottles, Burettes, Compact Burettes
Select a Size
$15.30
Estimated to ship onJune 01, 2025
Select a Size
About This Item
$15.30
Estimated to ship onJune 01, 2025
Recommended Products
material
low-density polyethylene
narrow-mouth bottle
round bottle
translucent bottle
feature
cap
manufacturer/tradename
BRAND 129055
capacity
500 mL
diam. × H
75 mm × 180 mm
volume
500 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service