Skip to Content
Merck
All Photos(1)

Key Documents

22837

Supelco

Desiccant Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
Pricing and availability is not currently available.

manufacturer/tradename

Parker Hannifin Corp. 1647727

Application

Desiccant cartridge may be incorporated in a reactor flow system to remove the water accumulated during conversion of benzyl alcohol to benzaldehyde.[1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalysis in flow: the practical and selective aerobic oxidation of alcohols to aldehydes and ketones.
Zotova, Natalia, et al.
Green Chemistry, 12, 2157-2163 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service