SLSV025LS
Millex®-SV Filter Unit (Sterile)
pore size 5.0 μm, diam. 25 mm, PVDF membrane, hydrophilic, medical device
Synonym(s):
Millex-SV Filter Unit, disposable syringe filter, sterile syringe filter, syringe filter
About This Item
Recommended Products
material
PVC housing
PVDF membrane
Quality Level
sterility
sterile; ethylene oxide treated
product line
Millex®
feature
CE compliant
holdup volume≤ 0.1 mL
hydrophilic
FDA registered
medical device
manufacturer/tradename
Millex®
parameter
≥600 mL/min water flow rate at 30 psi
45 °C max. temp.
5.2 bar max. inlet pressure (75 psi)
diam.
25 mm
filtration area
4 cm2
housing diam.
29 mm
inlet to outlet H
25 mm
volume
100 mL
matrix
Durapore®
pore size
5.0 μm pore size
fitting
female Luer-Lok® inlet
male Luer outlet slip
shipped in
ambient
General description
Application
- Filtration of aqueous, proteinaceous solutions where minimal sample loss is desired
- Particle or aggregate removal from protein solutions such as monoclonal antibodies, extractedcellular or tissue-based solutions, hormones, and drugs
Features and Benefits
- Single use only, non-pyrogenic and non-toxic
- 5.0 μm pore size Durapore® membrane filter sealed in a polyvinyl chloride (PVC) housing
- Compatible with most aqueous solutions
- CE marked and FDA registered
Other Notes
Legal Information
related product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service