Key Documents
BAF249760003
Workshields for β-emitters, free standing
W × L × H 31.4 cm (12.5 in.) × 40.6 cm (16 in.) × 52.1 cm (20.5 in.), size L
Select a Size
1 EA
$522.00
$522.00
Estimated to ship onMarch 29, 2025
Select a Size
Change View
1 EA
$522.00
About This Item
UNSPSC Code:
26142300
NACRES:
NB.35
$522.00
Estimated to ship onMarch 29, 2025
size
L
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F24976-0003
W × L × H
31.4 cm (12.5 in.) × 40.6 cm (16 in.) × 52.1 cm (20.5 in.)
General description
Select shield size to match requirements.
  • 9.5 mm (3/8 in.) thick, glass transparent acrylic shields protect against Beta sources and splashes from lab procedures
  • One piece construction has rounded corner for easy cleaning
  • Non skid feet prevent slipping on benchtop
  • Freestanding design has an angled shield and a flat base with a large work surface
Choose from one of the most recent versions:
Certificates of Analysis (COA)